Assay Details
| Gene Symbol: OTUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162403.1 | NP_001155875.1 | 3 - 4 | 242 | 69 | |
| GenBank mRNA | AK232154.1 | - | 3 - 4 | 239 | 69 | |
| AK392248.1 | - | 3 - 4 | 254 | 69 | ||
| AK398824.1 | - | 3 - 4 | 229 | 69 | ||
| AY609773.1 | - | 3 - 4 | 236 | 69 | ||
| EU310930.1 | - | 3 - 4 | 242 | 69 | ||
More Information
Set Membership: |
|

OTUB1
Genomic Map