Assay Details
| Gene Symbol: MSX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162887.1 | NP_001156359.1 | 1 - 2 | 469 | 78 | |
| GenBank mRNA | AB473487.1 | - | 1 - 2 | 469 | 78 | |
More Information
Set Membership: |
|

MSX1
Genomic Map