Assay Details
| Gene Symbol: LHX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170519.1 | NP_001163990.1 | 2 - 3 | 431 | 104 | |
| GenBank mRNA | AB473486.1 | - | 2 - 3 | 431 | 104 | |
More Information
Set Membership: |
|

LHX2
Genomic Map