Assay Details
| Gene Symbol: PROK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172586.1 | NP_001166057.1 | 1 - 2 | 69 | 60 | |
| GenBank mRNA | GU373677.1 | - | 1 - 2 | 69 | 60 | |
More Information
Set Membership: |
|

PROK1
Genomic Map