Assay Details
| Gene Symbol: SUN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177910.1 | NP_001171381.1 | 9 - 10 | 625 | 60 | |
| GenBank mRNA | GU475008.1 | - | 9 - 10 | 625 | 60 | |
More Information
Set Membership: |
|

SUN5
Genomic Map