Assay Details
| Gene Symbol: SPATA22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177921.1 | NP_001171392.1 | 7 - 8 | 907 | 74 | |
| XM_021066065.1 | XP_020921724.1 | 2007 | 74 | |||
| XM_021066066.1 | XP_020921725.1 | 2142 | 74 | |||
| XM_021066067.1 | XP_020921726.1 | 1206 | 74 | |||
| GenBank mRNA | GU474999.1 | - | 7 - 8 | 907 | 74 | |
More Information
Set Membership: |
|

SPATA22
Genomic Map