Assay Details
| Gene Symbol: PMM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184895.1 | NP_001171824.1 | 6 - 7 | 888 | 58 | |
| GenBank mRNA | AK235245.1 | - | 6 - 7 | 590 | 58 | |
| AK350347.1 | - | 6 - 7 | 589 | 58 | ||
| AK392413.1 | - | 6 - 7 | 593 | 58 | ||
| AK393586.1 | - | 6 - 7 | 595 | 58 | ||
More Information
Set Membership: |
|

PMM1
Genomic Map