Assay Details
| Gene Symbol: SELENOH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184948.1 | NP_001171877.1 | 1 - 2 | 136 | 73 | |
| GenBank mRNA | AK392739.1 | - | 1 - 2 | 326 | 73 | |
| HM018602.1 | - | 1 - 2 | 136 | 73 | ||
More Information
Set Membership: |
|

SELENOH
Genomic Map