Assay Details
Gene Symbol: RANBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185070.1 | NP_001171999.1 | 3 - 4 | 574 | 63 | |
XM_005670933.3 | XP_005670990.1 | 728 | 63 | |||
GenBank mRNA | AK237655.1 | - | 3 - 4 | 574 | 63 | |
AK239995.1 | - | 3 - 4 | 457 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
