Assay Details
| Gene Symbol: GADD45G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185129.1 | NP_001172058.1 | 1 - 2 | 146 | 90 | |
| GenBank mRNA | AK238667.1 | - | 1 - 2 | 160 | 90 | |
| GU386350.1 | - | 1 - 2 | 146 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001185129.1 transcripts.
Set Membership: |
|

GADD45G
Genomic Map