Assay Details
| Gene Symbol: GADD45G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185129.1 | NP_001172058.1 | 3 - 4 | 460 | 54 | |
| GenBank mRNA | GU386350.1 | - | 3 - 4 | 460 | 54 | |
More Information
Set Membership: |
|

GADD45G
Genomic Map