Assay Details
| Gene Symbol: FDPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185131.1 | NP_001172060.1 | 3 - 4 | 489 | 54 | |
| XM_005663310.3 | XP_005663367.1 | 415 | 54 | |||
| XM_021088556.1 | XP_020944215.1 | 548 | 54 | |||
| XM_021088557.1 | XP_020944216.1 | 554 | 54 | |||
| XM_021088559.1 | XP_020944218.1 | 415 | 54 | |||
| GenBank mRNA | AB529527.1 | - | 3 - 4 | 489 | 54 | |
| AK236486.1 | - | 4 - 5 | 710 | 54 | ||
| AK351226.1 | - | 3 - 4 | 331 | 54 | ||
| AK392898.1 | - | 3 - 4 | 416 | 54 | ||
| AK398138.1 | - | 3 - 4 | 416 | 54 | ||
| AK398787.1 | - | 4 - 5 | 548 | 54 | ||
| AY609787.1 | - | 3 - 4 | 734 | 54 | ||
More Information
Set Membership: |
|

FDPS
Genomic Map