Assay Details
| Gene Symbol: OAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185141.1 | NP_001172070.1 | 2 - 3 | 251 | 78 | |
| XM_005671546.3 | XP_005671603.1 | 432 | 78 | |||
| XM_005671548.3 | XP_005671605.2 | 376 | 78 | |||
| XM_021071999.1 | XP_020927658.1 | 497 | 78 | |||
| GenBank mRNA | AK231002.1 | - | 3 - 4 | 375 | 78 | |
| AK233181.1 | - | 3 - 4 | 373 | 78 | ||
| AK233676.1 | - | 2 - 3 | 290 | 78 | ||
| AK234316.1 | - | 2 - 3 | 285 | 78 | ||
| EU404089.1 | - | 2 - 3 | 251 | 78 | ||
More Information
Set Membership: |
|

OAT
Genomic Map