Assay Details
| Gene Symbol: SLA-DOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185143.1 | NP_001172072.1 | 1 - 2 | 125 | 83 | |
| GenBank mRNA | AB012858.1 | - | 1 - 2 | 111 | 83 | |
| AK231610.1 | - | 1 - 2 | 162 | 83 | ||
| AK347069.1 | - | 1 - 2 | 151 | 83 | ||
| KU705644.1 | - | 1 - 2 | 102 | 83 | ||
More Information
Set Membership: |
|

SLA-DOA
Genomic Map