Assay Details
| Gene Symbol: SLA-DOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185143.1 | NP_001172072.1 | 3 - 4 | 651 | 55 | |
| GenBank mRNA | AB012858.1 | - | 3 - 4 | 637 | 55 | |
| AK231610.1 | - | 3 - 4 | 688 | 55 | ||
| AK347069.1 | - | 3 - 4 | 677 | 55 | ||
| KU705644.1 | - | 3 - 4 | 628 | 55 | ||
More Information
Set Membership: |
|

SLA-DOA
Genomic Map