Assay Details
| Gene Symbol: DCSTAMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185150.1 | NP_001172079.1 | 1 - 2 | 35 | 57 | |
| XM_021088630.1 | XP_020944289.1 | 123 | 57 | |||
| GenBank mRNA | AK231004.1 | - | 1 - 2 | 35 | 57 | |
| AK389845.1 | - | 1 - 2 | 35 | 57 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001185150.1, XM_021088630.1 transcripts.
Set Membership: |
|

DCSTAMP
Genomic Map