Assay Details
| Gene Symbol: HMGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185154.1 | NP_001172083.1 | 4 - 5 | 431 | 97 | |
| NM_001185155.1 | NP_001172084.1 | 4 - 5 | 465 | 97 | ||
| XM_021098125.1 | XP_020953784.1 | 513 | 97 | |||
| XM_021098126.1 | XP_020953785.1 | 390 | 97 | |||
| GenBank mRNA | AK236172.1 | - | 4 - 5 | 434 | 97 | |
| AK238937.1 | - | 4 - 5 | 465 | 97 | ||
| AK396148.1 | - | 4 - 5 | 430 | 97 | ||
| AK398319.1 | - | 4 - 5 | 465 | 97 | ||
| EU559627.1 | - | 2 - 3 | 187 | 97 | ||
More Information
Set Membership: |
|

HMGA1
Genomic Map