Assay Details
| Gene Symbol: SCAMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185169.1 | NP_001172098.1 | 1 - 2 | 279 | 53 | |
| GenBank mRNA | AK236023.1 | - | 1 - 2 | 180 | 53 | |
| AK236431.1 | - | 1 - 2 | 269 | 53 | ||
| AK351549.1 | - | 273 | 53 | |||
| AK396133.1 | - | 1 - 2 | 279 | 53 | ||
More Information
Set Membership: |
|

SCAMP3
Genomic Map