Assay Details
| Gene Symbol: RGS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185184.1 | NP_001172113.1 | 3 - 4 | 333 | 79 | |
| GenBank mRNA | AK231087.1 | - | 3 - 4 | 333 | 79 | |
| AK391737.1 | - | 3 - 4 | 335 | 79 | ||
More Information
Set Membership: |
|

RGS4
Genomic Map