Assay Details
| Gene Symbol: APOA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001186294.1 | NP_001173223.1 | 1 - 2 | 35 | 59 | |
| GenBank mRNA | AK233612.1 | - | 1 - 2 | 35 | 59 | |
| AK345816.1 | - | 1 - 2 | 33 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001186294.1 transcripts.
Set Membership: |
|

APOA2
Genomic Map