Assay Details
Gene Symbol: CHCHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190155.1 | NP_001177084.1 | 2 - 3 | 274 | 70 | |
GenBank mRNA | AK236996.1 | - | 2 - 3 | 273 | 70 | |
AY610358.1 | - | 2 - 3 | 255 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
