Assay Details
| Gene Symbol: HOGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190169.1 | NP_001177098.1 | 3 - 4 | 821 | 61 | |
| GenBank mRNA | AK236562.1 | - | 3 - 4 | 821 | 61 | |
| AK393618.1 | - | 3 - 4 | 489 | 61 | ||
More Information
Set Membership: |
|

HOGA1
Genomic Map