Assay Details
| Gene Symbol: ECHS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190175.1 | NP_001177104.1 | 6 - 7 | 825 | 81 | |
| GenBank mRNA | AK232518.1 | - | 6 - 7 | 825 | 81 | |
| AK237861.1 | - | 6 - 7 | 1226 | 81 | ||
| AK346919.1 | - | 6 - 7 | 753 | 81 | ||
| AK389542.1 | - | 6 - 7 | 979 | 81 | ||
| AK392778.1 | - | 6 - 7 | 746 | 81 | ||
More Information
Set Membership: |
|

ECHS1
Genomic Map