Assay Details
| Gene Symbol: MTFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190176.1 | NP_001177105.1 | 2 - 3 | 447 | 83 | |
| XM_005654380.3 | XP_005654437.1 | 3074 | 83 | |||
| XM_013984860.2 | XP_013840314.1 | 3074 | 83 | |||
| GenBank mRNA | AK232190.1 | - | 2 - 3 | 447 | 83 | |
| AK345725.1 | - | 2 - 3 | 447 | 83 | ||
More Information
Set Membership: |
|

MTFR2
Genomic Map