Assay Details
| Gene Symbol: RPL30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190178.1 | NP_001177107.1 | 2 - 3 | 142 | 70 | |
| XM_021088585.1 | XP_020944244.1 | 187 | 70 | |||
| GenBank mRNA | AK239618.1 | - | 2 - 3 | 100 | 70 | |
| AK348689.1 | - | 2 - 3 | 96 | 70 | ||
| CB479993.1 | - | 3 - 4 | 370 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001190178.1, XM_021088585.1 transcripts.
Set Membership: |
|

RPL30
Genomic Map