Assay Details
| Gene Symbol: RPL30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190178.1 | NP_001177107.1 | 3 - 4 | 283 | 79 | |
| XM_021088585.1 | XP_020944244.1 | 328 | 79 | |||
| GenBank mRNA | AK234547.1 | - | 255 | 79 | ||
| AK239618.1 | - | 3 - 4 | 241 | 79 | ||
| AK348689.1 | - | 3 - 4 | 237 | 79 | ||
More Information
Set Membership: |
|

RPL30
Genomic Map