Assay Details
| Gene Symbol: RPL30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190178.1 | NP_001177107.1 | 4 - 5 | 417 | 92 | |
| XM_021088585.1 | XP_020944244.1 | 462 | 92 | |||
| GenBank mRNA | AK234547.1 | - | 389 | 92 | ||
| AK239618.1 | - | 4 - 5 | 375 | 92 | ||
| AK348689.1 | - | 4 - 5 | 371 | 92 | ||
| CB479993.1 | - | 1 - 2 | 95 | 92 | ||
More Information
Set Membership: |
|

RPL30
Genomic Map