Assay Details
| Gene Symbol: KRTCAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190186.1 | NP_001177115.1 | 3 - 4 | 300 | 70 | |
| GenBank mRNA | AK235351.1 | - | 3 - 4 | 304 | 70 | |
| AK238420.1 | - | 3 - 4 | 244 | 70 | ||
| AK400712.1 | - | 3 - 4 | 313 | 70 | ||
More Information
Set Membership: |
|

KRTCAP2
Genomic Map