Assay Details
| Gene Symbol: CAMK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190195.2 | NP_001177124.2 | 7 - 8 | 800 | 77 | |
| XM_021068250.1 | XP_020923909.1 | 830 | 77 | |||
| XM_021068251.1 | XP_020923910.1 | 661 | 77 | |||
| XM_021068252.1 | XP_020923911.1 | 638 | 77 | |||
| GenBank mRNA | AK237677.1 | - | 7 - 8 | 772 | 77 | |
More Information
Set Membership: |
|

CAMK1
Genomic Map