Assay Details
| Gene Symbol: COX6A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190221.1 | NP_001177150.1 | 1 - 2 | 138 | 72 | |
| GenBank mRNA | AJ679702.1 | - | 1 - 2 | 126 | 72 | |
| AK230744.1 | - | 1 - 2 | 116 | 72 | ||
| AK231185.1 | - | 1 - 2 | 138 | 72 | ||
| AK392232.1 | - | 1 - 2 | 148 | 72 | ||
| AK392351.1 | - | 1 - 2 | 118 | 72 | ||
| AY610231.1 | - | 1 - 2 | 62 | 72 | ||
| EW103555.2 | - | 1 - 2 | 67 | 72 | ||
More Information
Set Membership: |
|

COX6A1
Genomic Map