Assay Details
| Gene Symbol: GPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190240.1 | NP_001177169.1 | 7 - 8 | 1200 | 81 | |
| XM_021090918.1 | XP_020946577.1 | 1265 | 81 | |||
| GenBank mRNA | AK232843.1 | - | 7 - 8 | 1200 | 81 | |
More Information
Set Membership: |
|

GPD1
Genomic Map