Assay Details
| Gene Symbol: RXRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190246.1 | NP_001177175.1 | 4 - 5 | 821 | 94 | |
| XM_005665857.2 | XP_005665914.1 | 1009 | 94 | |||
| XM_005665858.3 | XP_005665915.1 | 616 | 94 | |||
| XM_021098096.1 | XP_020953755.1 | 616 | 94 | |||
| GenBank mRNA | DQ866835.1 | - | 4 - 5 | 821 | 94 | |
More Information
Set Membership: |
|

RXRB
Genomic Map