Assay Details
| Gene Symbol: GABARAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190288.1 | NP_001177217.1 | 3 - 4 | 391 | 102 | |
| GenBank mRNA | AK233704.1 | - | 3 - 4 | 391 | 102 | |
| AK392528.1 | - | 3 - 4 | 403 | 102 | ||
| AY610317.1 | - | 3 - 4 | 333 | 102 | ||
| HM046514.1 | - | 3 - 4 | 391 | 102 | ||
More Information
Set Membership: |
|

GABARAP
Genomic Map