Assay Details
| Gene Symbol: MAP1LC3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190290.1 | NP_001177219.1 | 2 - 3 | 221 | 65 | |
| GenBank mRNA | AK231642.1 | - | 2 - 3 | 198 | 65 | |
| AK389802.1 | - | 2 - 3 | 198 | 65 | ||
| AK392018.1 | - | 2 - 3 | 199 | 65 | ||
| HM046515.1 | - | 2 - 3 | 221 | 65 | ||
More Information
Set Membership: |
|

MAP1LC3B
Genomic Map