Assay Details
| Gene Symbol: PNLIPRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190291.1 | NP_001177220.1 | 6 - 7 | 786 | 57 | |
| XM_021072050.1 | XP_020927709.1 | 1122 | 57 | |||
| GenBank mRNA | AK231359.1 | - | 7 - 8 | 820 | 57 | |
| AK345102.1 | - | 7 - 8 | 805 | 57 | ||
| EU715062.1 | - | 6 - 7 | 786 | 57 | ||
More Information
Set Membership: |
|

PNLIPRP2
Genomic Map