Assay Details
| Gene Symbol: VIPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195117.1 | NP_001182046.1 | 8 - 9 | 861 | 58 | |
| GenBank mRNA | GU576975.1 | - | 8 - 9 | 861 | 58 | |
| KJ410050.1 | - | 9 - 10 | 967 | 58 | ||
More Information
Set Membership: |
|

VIPR2
Genomic Map