Assay Details
Gene Symbol: ADAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195323.1 | NP_001182252.1 | 6 - 7 | 759 | 68 | |
XM_005669057.3 | XP_005669114.1 | 716 | 68 | |||
XM_021066053.1 | XP_020921712.1 | 790 | 68 | |||
XM_021066054.1 | XP_020921713.1 | 1347 | 68 | |||
XM_021066055.1 | XP_020921714.1 | 1109 | 68 | |||
XM_021066056.1 | XP_020921715.1 | 716 | 68 | |||
XM_021066057.1 | XP_020921716.1 | 1216 | 68 | |||
XM_021066058.1 | XP_020921717.1 | 978 | 68 | |||
XM_021066059.1 | XP_020921718.1 | 585 | 68 | |||
XM_021066060.1 | XP_020921719.1 | 795 | 68 | |||
XM_021066061.1 | XP_020921720.1 | 790 | 68 | |||
XM_021066062.1 | XP_020921721.1 | 788 | 68 | |||
XM_021066063.1 | XP_020921722.1 | 801 | 68 | |||
GenBank mRNA | AK345566.1 | - | 6 - 7 | 763 | 68 | |
AK390102.1 | - | 7 - 8 | 697 | 68 | ||
GU373706.1 | - | 6 - 7 | 759 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
