Assay Details
| Gene Symbol: HOXA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195342.1 | NP_001182271.1 | 2 - 3 | 398 | 69 | |
| XM_021078465.1 | XP_020934124.1 | 1020 | 69 | |||
| GenBank mRNA | GU373678.1 | - | 2 - 3 | 398 | 69 | |
More Information
Set Membership: |
|

HOXA13
Genomic Map