Assay Details
| Gene Symbol: NPM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195362.1 | NP_001182291.1 | 4 - 5 | 381 | 74 | |
| XM_005657214.3 | XP_005657271.2 | 377 | 74 | |||
| XM_005657216.3 | XP_005657273.2 | 1255 | 74 | |||
| XM_021071934.1 | XP_020927593.1 | 1251 | 74 | |||
| XM_021071935.1 | XP_020927594.1 | 1134 | 74 | |||
| XM_021071936.1 | XP_020927595.1 | 1253 | 74 | |||
| GenBank mRNA | GU373718.1 | - | 4 - 5 | 381 | 74 | |
More Information
Set Membership: |
|

NPM2
Genomic Map