Assay Details
| Gene Symbol: MGARP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195363.2 | NP_001182292.1 | 2 - 3 | 222 | 72 | |
| GenBank mRNA | AK343386.1 | - | 2 - 3 | 221 | 72 | |
| AK348480.1 | - | 2 - 3 | 219 | 72 | ||
| GU373669.1 | - | 2 - 3 | 221 | 72 | ||
| HX212195.1 | - | 2 - 3 | 223 | 72 | ||
More Information
Set Membership: |
|

MGARP
Genomic Map