Assay Details
Gene Symbol: MGARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195363.2 | NP_001182292.1 | 3 - 4 | 315 | 87 | |
GenBank mRNA | AK343386.1 | - | 3 - 4 | 314 | 87 | |
AK348480.1 | - | 3 - 4 | 312 | 87 | ||
GU373669.1 | - | 3 - 4 | 314 | 87 | ||
HX212195.1 | - | 3 - 4 | 316 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
