Assay Details
| Gene Symbol: SDR16C5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195368.1 | NP_001182297.1 | 3 - 4 | 567 | 74 | |
| XM_013996752.2 | XP_013852206.1 | 846 | 74 | |||
| GenBank mRNA | AK344212.1 | - | 4 - 5 | 667 | 74 | |
| GU373688.1 | - | 3 - 4 | 567 | 74 | ||
More Information
Set Membership: |
|

SDR16C5
Genomic Map