Assay Details
| Gene Symbol: SMAD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195510.1 | NP_001182439.1 | 2 - 3 | 685 | 80 | |
| GenBank mRNA | AK395968.1 | - | 3 - 4 | 1008 | 80 | |
| HQ148186.1 | - | 2 - 3 | 685 | 80 | ||
More Information
Set Membership: |
|

SMAD9
Genomic Map