Assay Details
Gene Symbol: SEC14L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198918.1 | NP_001185847.1 | 8 - 9 | 915 | 66 | |
XM_005670808.2 | XP_005670865.1 | 1575 | 66 | |||
XM_005670810.3 | XP_005670867.1 | 1095 | 66 | |||
XM_005670811.3 | XP_005670868.1 | 1068 | 66 | |||
XM_005670815.3 | XP_005670872.1 | 1110 | 66 | |||
XM_013983047.2 | XP_013838501.1 | 1452 | 66 | |||
XM_021071866.1 | XP_020927525.1 | 1487 | 66 | |||
XM_021071867.1 | XP_020927526.1 | 1819 | 66 | |||
GenBank mRNA | AB530153.1 | - | 8 - 9 | 915 | 66 | |
AK233385.1 | - | 10 - 11 | 1205 | 66 | ||
AK394068.1 | - | 11 - 12 | 1129 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
