Assay Details
| Gene Symbol: PSAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198919.1 | NP_001185848.1 | 13 - 14 | 1555 | 65 | |
| XM_005671041.3 | XP_005671098.1 | 1659 | 65 | |||
| XM_005671042.3 | XP_005671099.1 | 1656 | 65 | |||
| XM_005671043.3 | XP_005671100.1 | 1656 | 65 | |||
| XM_021071888.1 | XP_020927547.1 | 1653 | 65 | |||
| XM_021071889.1 | XP_020927548.1 | 1647 | 65 | |||
| XM_021071890.1 | XP_020927549.1 | 1537 | 65 | |||
| XM_021071892.1 | XP_020927551.1 | 1603 | 65 | |||
| GenBank mRNA | HQ245644.1 | - | 14 - 15 | 1564 | 65 | |
| HQ245645.1 | - | 13 - 14 | 1555 | 65 | ||
More Information
Set Membership: |
|

PSAP
Genomic Map