Assay Details
| Gene Symbol: DHX58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199132.1 | NP_001186061.1 | 11 - 12 | 2003 | 63 | |
| XM_005668814.3 | XP_005668871.1 | 2002 | 63 | |||
| XM_005668815.3 | XP_005668872.1 | 1889 | 63 | |||
| GenBank mRNA | AK349545.1 | - | 10 - 11 | 1784 | 63 | |
| FJ392007.1 | - | 11 - 12 | 2003 | 63 | ||
More Information
Set Membership: |
|

DHX58
Genomic Map