Assay Details
| Gene Symbol: EIF4EBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202520.1 | NP_001189449.1 | 1 - 2 | 169 | 96 | |
| GenBank mRNA | EW449528.2 | - | 1 - 2 | 136 | 96 | |
| FD597014.1 | - | 1 - 2 | 169 | 96 | ||
More Information
Set Membership: |
|

EIF4EBP3
Genomic Map