Assay Details
| Gene Symbol: TICAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204351.1 | NP_001191280.1 | 1 | 2046 | 85 | |
| XM_021079196.1 | XP_020934855.1 | 2312 | 85 | |||
| XM_021079199.1 | XP_020934858.1 | 3043 | 85 | |||
| XM_021079209.1 | XP_020934868.1 | 2341 | 85 | |||
| GenBank mRNA | AJ947460.1 | - | 1 | 116 | 85 | |
More Information
Set Membership: |
|

TICAM2
Genomic Map