Assay Details
| Gene Symbol: C4H1orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204390.1 | NP_001191319.1 | 3 - 4 | 542 | 78 | |
| GenBank mRNA | AK233670.1 | - | 3 - 4 | 510 | 78 | |
| AK234116.1 | - | 3 - 4 | 510 | 78 | ||
| AK346595.1 | - | 3 - 4 | 510 | 78 | ||
| AK351176.1 | - | 3 - 4 | 437 | 78 | ||
| AK397935.1 | - | 3 - 4 | 585 | 78 | ||
| AY609895.1 | - | 3 - 4 | 435 | 78 | ||
| EU814887.1 | - | 3 - 4 | 511 | 78 | ||
More Information
Set Membership: |
|

C4H1orf43
Genomic Map