Assay Details
| Gene Symbol: C4H1orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204390.1 | NP_001191319.1 | 4 - 5 | 594 | 96 | |
| GenBank mRNA | AK233670.1 | - | 4 - 5 | 562 | 96 | |
| AK234116.1 | - | 4 - 5 | 562 | 96 | ||
| AK346595.1 | - | 4 - 5 | 562 | 96 | ||
| AK351176.1 | - | 4 - 5 | 489 | 96 | ||
| AK397935.1 | - | 4 - 5 | 637 | 96 | ||
| AY609895.1 | - | 4 - 5 | 487 | 96 | ||
| EU814887.1 | - | 4 - 5 | 563 | 96 | ||
More Information
Set Membership: |
|

C4H1orf43
Genomic Map